N-Benzylaminoacetaldehyde diethyl acetal
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FLUO018205-1GEA
0
GBP
FLUO018205-1G
FLUO018205-5G
N-Benzylaminoacetaldehyde diethyl acetal
N-Benzylaminoacetaldehyde diethyl acetal
Formula:
C₁₃H₂₁NO₂ MW: 223.32 g/mol |
MDL Number:
MFCD00051599 CAS Number: 61190-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...