4'-(3,4-Difluorophenoxy)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023246-5G 023246-25G
FLUO023246-5GEA 0 GBP
FLUO023246-5G FLUO023246-25G
4'-(3,4-Difluorophenoxy)acetophenone
4'-(3,4-Difluorophenoxy)acetophenone
Formula: C₁₄H₁₀F₂O₂
MDL Number: MFCD06411655
CAS Number: 845866-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR