4'-Amino-3',5'-dichloroacetophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4′-Amino-3′,5′-dichloroacetophenone
M-Clarity™ quality level = MQ100
SIAL536148-25GEA
105
GBP
SIAL536148-25G
4'-Amino-3',5'-dichloroacetophenone, Sigma-Aldrich®
4'-Amino-3',5'-dichloroacetophenone
Formula:
C₈H₇Cl₂NO MW: 204.0500 g/mol |
MDL Number:
MFCD00238535 CAS Number: 37148-48-4 EINECS: 253-368-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...