4'-Amino-3',5'-dichloroacetophenone

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS21995G25EA 0 GBP
ACTIAS21995G25 ACTIAS21995G100
4'-Amino-3',5'-dichloroacetophenone
4'-Amino-3',5'-dichloroacetophenone
Formula: C₈H₇Cl₂NO
MW: 204.0500 g/mol
MDL Number: MFCD00238535
CAS Number: 37148-48-4
EINECS: 253-368-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR