4-(Phenylazo)diphenylamine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Phenylazodiphenylamine

MOLEM23320388EA 0 GBP
MOLEM23320388
4-(Phenylazo)diphenylamine
4-(Phenylazo)diphenylamine
Formula: C₁₈H₁₅N₃
MW: 273.34 g/mol
MDL Number: MFCD00003023
CAS Number: 101-75-7
EINECS: 202-972-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR