5-Fluoro-2-formylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Fluoro-2-formylphenyl)boronic acid

BB-9055-5G BB-9055-1G
COBBBB-9055-5GEA 0 GBP
COBBBB-9055-5G COBBBB-9055-1G
5-Fluoro-2-formylphenylboronic acid
5-Fluoro-2-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167.9300 g/mol
MDL Number: MFCD09953731
CAS Number: 1256355-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR