3-Fluoro-4-formylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-4-formylbenzeneboronic acid

BB-3474-25G BB-3474-1G BB-3474-5G
COBBBB-3474-25GEA 0 GBP
COBBBB-3474-25G COBBBB-3474-1G COBBBB-3474-5G
3-Fluoro-4-formylphenylboronic acid
3-Fluoro-4-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167.93 g/mol
MDL Number: MFCD02093051
CAS Number: 248270-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR