N-Butyl 4-boronobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(n-Butylsulphamoyl)benzeneboronic acid

BB-2428-1G BB-2428-5G
COBBBB-2428-1GEA 0 GBP
COBBBB-2428-1G COBBBB-2428-5G
N-Butyl 4-boronobenzenesulphonamide
N-Butyl 4-boronobenzenesulphonamide
Formula: C₁₀H₁₆BNO₄S
MW: 257.11 g/mol
MDL Number: MFCD07363753
CAS Number: 850589-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR