4-Boronobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Sulphamoylbenzeneboronic acid, (4-Aminosulphonylphenyl)boronic acid

BB-2402-5G BB-2402-1G
COBBBB-2402-5GEA 0 GBP
COBBBB-2402-5G COBBBB-2402-1G
4-Boronobenzenesulphonamide
4-Boronobenzenesulphonamide
Formula: C₆H₈BNO₄S
MW: 201.00 g/mol
MDL Number: MFCD06659839
CAS Number: 613660-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR