Hesperetin

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4'-Methoxy-3',5,7-trihydroxyflavanone(S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one(±)-Hesperetin3',5,7-Trihydroxy-4'-methoxyflavanone

ROTH7743.1EA 62.78 GBP
ROTH7743.1
Hesperetin
Hesperetin
Formula: C₁₆H₁₄O₆
MW: 302.30 g/mol
MDL Number: MFCD00075646
CAS Number: 520-33-2
EINECS: 208-290-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR