2-Methoxy-5-pyrimidineboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Methoxypyrimidin-5-yl)boronic acid

016194-1G 016194-5G
FLUO016194-1GEA 0 GBP
FLUO016194-1G FLUO016194-5G
2-Methoxy-5-pyrimidineboronic acid
2-Methoxy-5-pyrimidineboronic acid
Formula: C₅H₇BN₂O₃
MW: 153.9317 g/mol
MDL Number: MFCD03094664
CAS Number: 628692-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR