2-Methoxy-5-pyrimidineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Methoxypyrimidin-5-yl)boronic acid

BB-8374-5G BB-8374-1G BB-8374-25G
COBBBB-8374-5GEA 0 GBP
COBBBB-8374-5G COBBBB-8374-1G COBBBB-8374-25G
2-Methoxy-5-pyrimidineboronic acid
2-Methoxy-5-pyrimidineboronic acid
Formula: C₅H₇BN₂O₃
MW: 153.9317 g/mol
MDL Number: MFCD03094664
CAS Number: 628692-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR