2-Methoxy-5-pyrimidineboronic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Methoxypyrimidin-5-yl)boronic acid
AS50113G1
AS50113G25
AS50113G5
ACTIAS50113G1EA
0
GBP
ACTIAS50113G1
ACTIAS50113G25
ACTIAS50113G5
2-Methoxy-5-pyrimidineboronic acid
2-Methoxy-5-pyrimidineboronic acid
Formula:
C₅H₇BN₂O₃ MW: 153.9317 g/mol |
MDL Number:
MFCD03094664 CAS Number: 628692-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...