5-Pyrimidineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Pyrimidine-5-boronic acid, 5-Pyrimidylboronic acid, Pyrimidin-5-ylboronic acid, 5-Pyrimidinylboronic acid
BB-2360-5G
BB-2360-25G
BB-2360-1G
BB-2360-100G
COBBBB-2360-5GEA
0
GBP
COBBBB-2360-5G
COBBBB-2360-25G
COBBBB-2360-1G
COBBBB-2360-100G
5-Pyrimidineboronic acid
5-Pyrimidineboronic acid
Formula:
C₄H₅BN₂O₂ MW: 123.91 g/mol |
MDL Number:
MFCD03002366 CAS Number: 109299-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...