4-Iodo-3-methyl-1H-pyrazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-8020-5G
OT-8020-1G
COBBOT-8020-5GEA
0
GBP
COBBOT-8020-5G
COBBOT-8020-1G
4-Iodo-3-methyl-1H-pyrazole
4-Iodo-3-methyl-1H-pyrazole
Formula:
C₄H₅IN₂ MW: 208.0000 g/mol |
MDL Number:
MFCD09908176 CAS Number: 15802-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...