5-Amino-4-bromo-1-methyl-1H-pyrazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-1-methyl-1H-pyrazol-5-amine

CFF537.1 005503-1G
CHMPCFF537.1EA 0 GBP
CHMPCFF537.1 FLUO005503-1G
5-Amino-4-bromo-1-methyl-1H-pyrazole
5-Amino-4-bromo-1-methyl-1H-pyrazole
Formula: C₄H₆BrN₃
MW: 176.02 g/mol
MDL Number: MFCD00068157
CAS Number: 105675-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR