8,10-Heneicosadiynoic acid
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FLUO009256-5GEA
0
GBP
FLUO009256-5G
FLUO009256-1G
8,10-Heneicosadiynoic acid
8,10-Heneicosadiynoic acid
Formula:
C₂₁H₃₄O₂ |
MDL Number:
MFCD00190226 CAS Number: 174063-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...