(±)-3-Amino-1-boc-piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

AM-1606-5G AM-1606-1G AM-1606-25G
COBBAM-1606-5GEA 0 GBP
COBBAM-1606-5G COBBAM-1606-1G COBBAM-1606-25G
(±)-3-Amino-1-boc-piperidine
3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD01861219
CAS Number: 184637-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR