4-Amino-1-boc-piperidine-4-carboxylic acid
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid
BACE4027964.0005EA
1140
GBP
BACE4027964.0005
BACE4027964.0025
BACE4027964.0001
4-Amino-1-boc-piperidine-4-carboxylic acid
4-Amino-1-Boc-piperidine-4-carboxylic acid
Formula:
C₁₁H₂₀N₂O₄ MW: 244.29 g/mol |
MDL Number:
MFCD01318728 CAS Number: 183673-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...