Ionomycin calcium salt

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SERA26379.01EA 0 GBP
SERA26379.01
Ionomycin calcium salt
Ionomycin calcium salt
Formula: C₄₁H₇₀CaO₉
MDL Number: MFCD00151183
CAS Number: 56092-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR