Strychnine hemisulphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

S7001-100G S7001-25G S7001-5G
SIALS7001-100GEA 638 GBP
SIALS7001-100G SIALS7001-25G SIALS7001-5G
Strychnine hemisulphate, Sigma-Aldrich®
Strychnine hemisulphate
Formula: C₄₂H₄₆N₄O₈S
MDL Number: MFCD00068328
CAS Number: 60-41-3
EINECS: 200-477-7
UN: 1692
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR