Retinoic acid

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: all-trans-Retinoic acidVitamin A acidATRATretinoin

EHERC17608000EA 130 GBP
EHERC17608000
Retinoic acid
Retinoic acid
Formula: C₂₀H₂₈O₂
MW: 300,44
MDL Number: MFCD00001551
CAS Number: 302-79-4
EINECS: 206-129-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR