Folic acid
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Pteroyl-L-glutamic acid
, Vitamin B9
, PteGlu
, Vitamin M
, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid
SERA21700.02EA
27.94
GBP
SERA21700.02
SERA21700.03
Folic acid
Folic acid
Formula:
C₁₉H₁₉N₇O₆ MW: 441.41 g/mol |
MDL Number:
MFCD00079305 CAS Number: 59-30-3 EINECS: 200-419-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...