Tetrahydropyran-4-acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tetrahydropyranyl-4-acetic acid

AS12603G1 AS12603G25 AS12603G5
ACTIAS12603G1EA 0 GBP
ACTIAS12603G1 ACTIAS12603G25 ACTIAS12603G5
Tetrahydropyran-4-acetic acid
Tetrahydropyran-4-acetic acid
Formula: C₇H₁₂O₃
MW: 144.1700 g/mol
MDL Number: MFCD01631204
CAS Number: 85064-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR