N(α)-Boc-D-histidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Boc-D-His-OH

SIAL15119-1GEA 120 GBP
SIAL15119-1G
N(α)-Boc-D-histidine, Sigma-Aldrich®
N(α)-Boc-D-histidine
Formula: C₁₁H₁₇N₃O₄
MW: 255.3 g/mol
MDL Number: MFCD00037851
CAS Number: 50654-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR