[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex

AS20423G1 AS20423G5
ACTIAS20423G1EA 49 GBP
ACTIAS20423G1 ACTIAS20423G5
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride dichloromethane complex
Formula: C₃₅H₃₀Cl₄FeP₂Pd
MW: 816.64 g/mol
MDL Number: MFCD00792899
CAS Number: 95464-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR