4-Methoxyphenyl chloroformate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxyphenyl carbonochloridate

M-Clarity™ quality level = MQ100

322393-25G 322393-5G
SIAL322393-25GEA 348 GBP
SIAL322393-25G SIAL322393-5G
4-Methoxyphenyl chloroformate, Sigma-Aldrich®
4-Methoxyphenyl chloroformate
Formula: C₇H₄BrClO₂
MW: 186.5924 g/mol
MDL Number: MFCD00013258
CAS Number: 7693-41-6
UN: 3277
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR