4-Chlorophenyl chloroformate

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

31175.18EA 0 GBP
31175.18
4-Chlorophenyl chloroformate
4-Chlorophenyl chloroformate
Formula: C₇H₄Cl₂O₂
Density: 1.365
Flash Pt: 103 °C (218 °F)
MDL Number: MFCD00000638
CAS Number: 7693-45-0
UN: 3277
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR