(+)-N,N,N',N'-Tetramethyl-L-tartardiamide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2R,3R)-N,N,N',N'-Tetramethyl-L-tartaric acid diamide
010852-20G
010852-50G
FLUO010852-20GEA
0
GBP
FLUO010852-20G
FLUO010852-50G
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide
Formula:
C₈H₁₆N₂O₄ MW: 204.2236 g/mol |
MDL Number:
MFCD00025672 CAS Number: 26549-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...