Methyl acetopyruvate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2,4-dioxopentanoate

M-Clarity™ quality level = MQ100

699675-25G 699675-5G
SIAL699675-25GEA 220 GBP
SIAL699675-25G SIAL699675-5G
Methyl acetopyruvate, Sigma-Aldrich®
Methyl acetopyruvate
Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00043638
CAS Number: 20577-61-1
EINECS: 243-891-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR