4-(1H-Pyrazol-1-yl)benzaldehyde
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3T-0309.500
3T-0309.1
3T-0309.5
BIOT3T-0309.500EA
0
GBP
BIOT3T-0309.500
BIOT3T-0309.1
BIOT3T-0309.5
4-(1H-Pyrazol-1-yl)benzaldehyde
4-(1H-Pyrazol-1-yl)benzaldehyde
Formula:
C₁₀H₈N₂O MW: 172.1833 g/mol |
MDL Number:
MFCD02681938 CAS Number: 99662-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...