4-(1H-Pyrazol-1-yl)benzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20340G25
AS20340G5
ACTIAS20340G25EA
148
GBP
ACTIAS20340G25
ACTIAS20340G5
4-(1H-Pyrazol-1-yl)benzaldehyde
4-(1H-Pyrazol-1-yl)benzaldehyde
Formula:
C₁₀H₈N₂O MW: 172.1833 g/mol |
MDL Number:
MFCD02681938 CAS Number: 99662-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...