2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Fluoro-3-(trifluoromethyl)phenacyl bromide, 2-Bromo-1-(4-fluoro-3-(trifluoromethyl)phenyl)ethanone
AS23916G100
AS23916G25
AS23916G5
ACTIAS23916G100EA
0
GBP
ACTIAS23916G100
ACTIAS23916G25
ACTIAS23916G5
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone
Formula:
C₉H₅BrF₄O MW: 285.0330 g/mol |
MDL Number:
MFCD03094509 CAS Number: 537050-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...