4-Chloro-2-fluoro-6-iodoaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS33230G1 AS33230G10 AS33230G5
ACTIAS33230G1EA 0 GBP
ACTIAS33230G1 ACTIAS33230G10 ACTIAS33230G5
4-Chloro-2-fluoro-6-iodoaniline
4-Chloro-2-fluoro-6-iodoaniline
Formula: C₆H₄ClFIN
MW: 271.46 g/mol
MDL Number: MFCD01318122
CAS Number: 216393-67-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR