4'-Hydroxybutyrophenone
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS25288G100
AS25288G25
ACTIAS25288G100EA
123
GBP
ACTIAS25288G100
ACTIAS25288G25
4'-Hydroxybutyrophenone
4'-Hydroxybutyrophenone
Formula:
C₁₀H₁₂O₂ |
MDL Number:
MFCD00020173 CAS Number: 1009-11-6 EINECS: 213-766-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...