D-tert-Leucine (H-D-Tle-OH)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-D-Tle-OH

AS21846G1 AS21846G5
ACTIAS21846G1EA 14 GBP
ACTIAS21846G1 ACTIAS21846G5
D-tert-Leucine (H-D-Tle-OH)
D-tert-Leucine (H-D-Tle-OH)
Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
MDL Number: MFCD00004265
CAS Number: 26782-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR