2,6-Dichloro-3-iodopyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS4037G1 AS4037G25 AS4037G5
ACTIAS4037G1EA 0 GBP
ACTIAS4037G1 ACTIAS4037G25 ACTIAS4037G5
2,6-Dichloro-3-iodopyridine
2,6-Dichloro-3-iodopyridine
Formula: C₅H₂Cl₂IN
MW: 273.88 g/mol
MDL Number: MFCD03094686
CAS Number: 148493-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR