4-(1H-1,2,4-Triazol-1-yl)benzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(4-Formylphenyl)-1,2,4-triazole, 4-(1,2,4-Triazol-1-yl)benzaldehyde

AS20702G25 AS20702G5
ACTIAS20702G25EA 156 GBP
ACTIAS20702G25 ACTIAS20702G5
4-(1H-1,2,4-Triazol-1-yl)benzaldehyde
4-(1H-1,2,4-Triazol-1-yl)benzaldehyde
Formula: C₉H₇N₃O
MW: 173.1714 g/mol
MDL Number: MFCD02681969
CAS Number: 27996-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR