3-(1H-1,2,4-Triazol-1-yl)benzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(1,2,4-Triazol-1-yl)benzaldehyde

AS20623G1 AS20623G5
ACTIAS20623G1EA 145 GBP
ACTIAS20623G1 ACTIAS20623G5
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde
Formula: C₉H₇N₃O
MW: 173.1714 g/mol
MDL Number: MFCD08271896
CAS Number: 868755-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR