N-Boc-D-prolinol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, N-tert-Butoxycarbonyl-D-prolinol, N-t-Boc-D-prolinol, Boc-D-Prolinol
AS24428G100
AS24428G25
AS24428G5
ACTIAS24428G100EA
83
GBP
ACTIAS24428G100
ACTIAS24428G25
ACTIAS24428G5
N-Boc-D-prolinol
N-Boc-D-prolinol
Formula:
C₁₀H₁₉NO₃ MW: 201.3 g/mol |
MDL Number:
MFCD00040580 CAS Number: 83435-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...