2-Chloro-3-fluoro-5-hydroxypyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS31094G1
AS31094G5
ACTIAS31094G1EA
0
GBP
ACTIAS31094G1
ACTIAS31094G5
2-Chloro-3-fluoro-5-hydroxypyridine
2-Chloro-3-fluoro-5-hydroxypyridine
Formula:
C₆H₄ClFO MW: 147.5400 g/mol |
MDL Number:
MFCD03411573 CAS Number: 870062-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...