tert-Butyl methyl malonate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert.-Butyl methyl malonate

AS21613G100 AS21613G25 AS21613G5
ACTIAS21613G100EA 155 GBP
ACTIAS21613G100 ACTIAS21613G25 ACTIAS21613G5
tert-Butyl methyl malonate
tert-Butyl methyl malonate
Formula: C₈H₁₄O₄
MW: 174.19 g/mol
MDL Number: MFCD00042856
CAS Number: 42726-73-8
EINECS: 255-919-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR