(±)-3-Hydroxytetrahydrofuran

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20747G100 AS20747G25
ACTIAS20747G100EA 83 GBP
ACTIAS20747G100 ACTIAS20747G25
(±)-3-Hydroxytetrahydrofuran
(±)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
MDL Number: MFCD00005374
CAS Number: 453-20-3
EINECS: 207-219-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR