2-Chloro-5-methylpyrazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS31679G1 AS31679G5
ACTIAS31679G1EA 0 GBP
ACTIAS31679G1 ACTIAS31679G5
2-Chloro-5-methylpyrazine
2-Chloro-5-methylpyrazine
Formula: C₅H₅ClN₂
MW: 128.5600 g/mol
MDL Number: MFCD00055039
CAS Number: 59303-10-5
EINECS: 202-434-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR