Methyl oxindole-5-carboxylate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl Oxindole-5-carboxylate
AS21290G1
AS21290G25
AS21290G5
ACTIAS21290G1EA
55
GBP
ACTIAS21290G1
ACTIAS21290G25
ACTIAS21290G5
Methyl oxindole-5-carboxylate
Methyl oxindole-5-carboxylate
Formula:
C₁₀H₉NO₃ MW: 191.1834 g/mol |
MDL Number:
MFCD02179600 CAS Number: 199328-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...