4-Chloro-5,7-dimethylquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21109G1 AS21109G5
ACTIAS21109G1EA 435 GBP
ACTIAS21109G1 ACTIAS21109G5
4-Chloro-5,7-dimethylquinoline
4-Chloro-5,7-dimethylquinoline
Formula: C₁₁H₁₀ClN
CAS Number: 352205-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR