4,6-Dichloro-5-methoxypyrimidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS50352G25
AS50352G5
ACTIAS50352G25EA
0
GBP
ACTIAS50352G25
ACTIAS50352G5
4,6-Dichloro-5-methoxypyrimidine
4,6-Dichloro-5-methoxypyrimidine
Formula:
C₅H₄Cl₂N₂O MW: 179.01 g/mol |
MDL Number:
MFCD00087680 CAS Number: 5018-38-2 EINECS: 225-699-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...