4-Chlorodiphenylmethane

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24904G100 AS24904G25
ACTIAS24904G100EA 72 GBP
ACTIAS24904G100 ACTIAS24904G25
4-Chlorodiphenylmethane
4-Chlorodiphenylmethane
Formula: C₁₃H₁₁Cl
MDL Number: MFCD00094070
CAS Number: 831-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR