2-Amino-6-chlorophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS34889G1 AS34889G5
ACTIAS34889G1EA 0 GBP
ACTIAS34889G1 ACTIAS34889G5
2-Amino-6-chlorophenol
2-Amino-6-chlorophenol
Formula: C₆H₆ClNO
MW: 143.5700 g/mol
MDL Number: MFCD07801210
CAS Number: 38191-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR