4-Quinolineboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinolin-4-ylboronic acid

AS21098G1 AS21098G5
ACTIAS21098G1EA 83 GBP
ACTIAS21098G1 ACTIAS21098G5
4-Quinolineboronic acid
4-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.9763 g/mol
MDL Number: MFCD03095174
CAS Number: 371764-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR